1,2,3-Tri-O-acetyl-5-deoxy-β-D-ribofuranose ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
(2S,3R,4R,5R)-5-Methyltetrahydrofuran-2,3,4-triyl triacetate
T2607-25G
T2607-5G
TCIAT2607-25GEA
152
EUR
TCIAT2607-25G
TCIAT2607-5G
1,2,3-Tri-O-acetyl-5-deoxy-β-D-ribofuranose ≥98.0% (by GC)
1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose
Formula:
C₁₁H₁₆O₇ Peso molecolare: 260,24 g/mol |
MDL Number:
MFCD08458459 CAS Number: 62211-93-2 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Specific rotation [a]20/D | -19.0 to -23.0 deg (C=2, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...