Stampa…

2,3,3-Trimethyl-3H-benzo[g]indole ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 2,3,3-Trimethylnaphtho[1,2-b]pyrrole

T2601-25G T2601-5G
TCIAT2601-25GEA 365 EUR
TCIAT2601-25G TCIAT2601-5G
2,3,3-Trimethyl-3H-benzo[g]indole ≥98.0% (by HPLC, titration analysis)
2,3,3-Trimethyl-3H-benzo[g]indole
Formula: C₁₅H₁₅N
Peso molecolare: 209,29 g/mol
Punto di ebollizione: 127°C
MDL Number: MFCD00723378
CAS Number: 74470-85-2

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR