1-[2-(Trimethylsilyl)ethoxycarbonyloxy]benzotriazole ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
Teoc-OBt
T2590-1G
T2590-5G
TCIAT2590-1GEA
125
EUR
TCIAT2590-1G
TCIAT2590-5G
1-[2-(Trimethylsilyl)ethoxycarbonyloxy]benzotriazole ≥98.0% (by HPLC)
1-[2-(Trimethylsilyl)ethoxycarbonyloxy]benzotriazole
Formula:
C₁₂H₁₇N₃O₃Si Peso molecolare: 279,37 g/mol |
CAS Number:
113306-55-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 14.30 to 15.80 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...