Stampa…

(1S,4S,5S)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane ≥98.0% (by GC)

Fornitore: TCI

T2579-1G T2579-5G
TCIAT2579-1GEA 89 EUR
TCIAT2579-1G TCIAT2579-5G
(1S,4S,5S)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane ≥98.0% (by GC)
(1S,4S,5S)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane
Formula: C₁₀H₁₈S
CAS Number: 1208985-45-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 97.0 ee%
Specific rotation [a]20/D +65.0 to +69.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR