Stampa…

2,3-Dihydro-1H-carbazol-4(9H)-one ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ondansetron Impurity I, 1,2,3,4-Tetrahydro-4-oxocarbazole, 1,2,3,4-Tetrahydrocarbazol-4-one

T2559-25G T2559-5G
TCIAT2559-25GEA 56 EUR
TCIAT2559-25G TCIAT2559-5G
2,3-Dihydro-1H-carbazol-4(9H)-one ≥98.0% (by GC)
2,3-Dihydro-1H-carbazol-4(9H)-one
Formula: C₁₂H₁₁NO
Peso molecolare: 185,2218 g/mol
MDL Number: MFCD00173749
CAS Number: 15128-52-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 223.0 to 229.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR