Stampa…

2,4,6-Trimethyl-2,4,6-trivinylcyclotrisilazane ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

T2556-25G T2556-5G
TCIAT2556-25GEA 158 EUR
TCIAT2556-25G TCIAT2556-5G
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisilazane ≥97.0% (by titrimetric analysis)
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisilazane
Formula: C₉H₂₁N₃Si₃
Peso molecolare: 255,54 g/mol
Punto di ebollizione: 65°C
Flash Pt: 85°C
MDL Number: MFCD00054934
CAS Number: 5505-72-6
EINECS: 226-848-3
UN: 3267
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 93.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR