Stampa…

Torsemide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Torasemide, N-(Isopropylcarbamoyl)-4-(m-tolylamino)pyridine-3-sulphonamide

T2538-1G T2538-5G
TCIAT2538-1GEA 59 EUR
TCIAT2538-1G TCIAT2538-5G
Torsemide ≥98.0% (by HPLC, titration analysis)
Torsemide
Formula: C₁₆H₂₀N₄O₃S
Peso molecolare: 348,42 g/mol
MDL Number: MFCD00866166
CAS Number: 56211-40-6
Merck Index: 14,09552

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR