Stampa…

5-Pyrimidineboronic acid pinacol ester ≥96.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

T2521-1G T2521-5G
TCIAT2521-1GEA 38 EUR
TCIAT2521-1G TCIAT2521-5G
5-Pyrimidineboronic acid pinacol ester ≥96.0% (by GC, titration analysis)
5-Pyrimidineboronic acid pinacol ester
Formula: C₁₀H₁₅BN₂O₂
MDL Number: MFCD05155223
CAS Number: 321724-19-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR