Thieno[3,2-b]thiophene ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
1,4-Dithiapentalene
T2513-1G
T2513-5G
TCIAT2513-1GEA
86
EUR
TCIAT2513-1G
TCIAT2513-5G
Thieno[3,2-b]thiophene ≥98.0% (by GC)
Thieno[3,2-b]thiophene
Formula:
C₆H₄S₂ Peso molecolare: 140,23 g/mol Punto di ebollizione: 71°C |
MDL Number:
MFCD00179479 CAS Number: 251-41-2 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 54.0 to 58.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...