Trifluorothymidine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
Trifluridine, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione, Trifluoromethyldeoxyuridine, 5-Trifluorothymidine
T2511-1G
T2511-100MG
TCIAT2511-1GEA
515
EUR
TCIAT2511-1G
TCIAT2511-100MG
Trifluorothymidine ≥98.0% (by HPLC, titration analysis)
Trifluorothymidine
Formula:
C₁₀H₁₁F₃N₂O₅ Peso molecolare: 296,2 g/mol Punto di fusione: 189°C |
MDL Number:
MFCD00006534 CAS Number: 70-00-8 EINECS: 200-722-8 Merck Index: 14,09687 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +47.0 to +53.0 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...