Tamoxifen citrate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
(Z)-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate
Formula:
C₂₆H₂₉NO Punto di fusione: 142°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00058321 CAS Number: 54965-24-1 EINECS: 259-415-2 Merck Index: 14,09048 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...