Stampa…

N-(Triphenylmethyl)-DL-serine methyl ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI

Synonyms: N-(Triphenylmethyl)-DL-serine Methyl Ester, N-Trityl-DL-serine Methyl Ester, Trt-DL-Ser-OMe, Methyl 3-hydroxy-2-(tritylamino)propanoate

T2500-25G T2500-5G
TCIAT2500-25GEA 218 EUR
TCIAT2500-25G TCIAT2500-5G
N-(Triphenylmethyl)-DL-serine methyl ester ≥98.0% (by GC, titration analysis)
N-(Triphenylmethyl)-DL-serine methyl ester
Formula: C₂₃H₂₃NO₃
MDL Number: MFCD00583950
CAS Number: 13515-76-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR