2,4,6-Trivinylboroxin pyridine complex ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Trivinylcyclotriboroxan - Pyridine Complex
T2498-1G
T2498-5G
TCIAT2498-1GEA
66
EUR
TCIAT2498-1G
TCIAT2498-5G
2,4,6-Trivinylboroxin pyridine complex ≥98.0% (by titrimetric analysis)
2,4,6-Trivinylboroxin pyridine complex
Formula:
C₁₁H₁₄B₃NO₃ Peso molecolare: 240,67 g/mol |
MDL Number:
MFCD03839440 CAS Number: 95010-17-6 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...