Stampa…

4-Thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Thiazole-4-carboxylic acid

T2494-1G T2494-5G
TCIAT2494-1GEA 53 EUR
TCIAT2494-1G TCIAT2494-5G
4-Thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)
4-Thiazolecarboxylic acid
Formula: C₄H₃NO₂S
Peso molecolare: 129,1371 g/mol
MDL Number: MFCD00623589
CAS Number: 3973-08-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4121e731051b426cfceca676f0ed2c9c3f7f1ea4&vid=4121e731051b426cfceca676f0ed2c9c3f7f1ea4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR