Stampa…

Tetrahydropyran-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tetrahydropyran-4-yl-carboxylic acid

T2493-5G T2493-25G
TCIAT2493-5GEA 30 EUR
TCIAT2493-5G TCIAT2493-25G
Tetrahydropyran-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Tetrahydropyran-4-carboxylic acid
Formula: C₆H₁₀O₃
Peso molecolare: 130,14 g/mol
Punto di ebollizione: 115°C
MDL Number: MFCD00031016
CAS Number: 5337-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8146f94a76989a8dc7974897179d22932be5d1d&vid=b8146f94a76989a8dc7974897179d22932be5d1d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 87.0 to 91.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR