Stampa…

2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl-2,2,2-trichloroacetimidate ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE

Synonyms: 2,3,4,6-Tetra-O-acetyl-1-O-trichloroacetimidoyl-beta-D-glucopyranoside, 2,3,4,6-Tetra-O-acetyl-b-D-glucopyranosyl trichloroacetimidate

T2491-1G T2491-5G
TCIAT2491-1GEA 110 EUR
TCIAT2491-1G TCIAT2491-5G
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl-2,2,2-trichloroacetimidate ≥98.0% (by total nitrogen basis)
2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl-2,2,2-trichloroacetimidate
Formula: C₁₆H₂₀Cl₃NO₁₀
Peso molecolare: 492,68 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD11112189
CAS Number: 92052-29-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 154.0 to 160.0 deg-C
Specific rotation [a]20/D +6.0 to +9.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR