Stampa…

1H,1H-Tridecafluoroheptyl iodide ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1H,1H-Perfluoro-1-iodoheptane, 1H,1H-Tridecafluoro-1-iodoheptane, 1H,1H-Perfluoroheptyl Iodide

T2482-1G T2482-5G
TCIAT2482-1GEA 58 EUR
TCIAT2482-1G TCIAT2482-5G
1H,1H-Tridecafluoroheptyl iodide ≥95.0% (by GC)
1H,1H-Tridecafluoroheptyl iodide
Formula: C₇H₂F₁₃I
Peso molecolare: 459,98 g/mol
MDL Number: MFCD00233026
CAS Number: 212563-43-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR