Trichostatin A ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
TSA, (R,2E,4E)-7-(4-(dimethylamino)phenyl)-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide, [R-(E,E)]-7-[4-(Dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide
T2477-10MG
T2477-50MG
TCIAT2477-10MGEA
306
EUR
TCIAT2477-10MG
TCIAT2477-50MG
Trichostatin A ≥98.0% (by HPLC)
Trichostatin A
Formula:
C₁₇H₂₂N₂O₃ Peso molecolare: 302,37 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD03848392 CAS Number: 58880-19-6 Merck Index: 14,09649 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 95.0 ee% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...