trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
trans-4-(Trifluoromethyl)cyclohexane-carboxylicacid
Formula:
C₈H₁₁F₃O₂ |
MDL Number:
MFCD09037363 CAS Number: 133261-33-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 151.0 to 155.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...