Stampa…

N-(Trimethylsilyl)bis(trifluoromethanesulphonyl)imide ≥97.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: TMS Triflimide, [Bis(trifluoromethanesulphonyl)amino]trimethylsilane

T2392-1G T2392-5G
TCIAT2392-1GEA 69 EUR
TCIAT2392-1G TCIAT2392-5G
N-(Trimethylsilyl)bis(trifluoromethanesulphonyl)imide ≥97.0% (by titrimetric analysis)
N-(Trimethylsilyl)bis(trifluoromethanesulphonyl)imide
Formula: C₅H₉F₆NO₄S₂Si
Punto di ebollizione: 115°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD10566930
CAS Number: 82113-66-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Red to Green clear liquid
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR