Trioxsalen ≥98.0% (by HPLC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
4,5',8-Trimethylpsoralen, Trisoralen, TMP, 2,5,9-Trimethylfuro[3,2-g]benzopyran-7-one
Formula:
C₁₄H₁₂O₃ Punto di fusione: 234°C |
MDL Number:
MFCD00005010 CAS Number: 3902-71-4 EINECS: 223-459-0 UN: 1759 ADR: 8,III Merck Index: 14,09735 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...