Stampa…

5,10,15,20-Tetra(4-pyridyl)porphyrin ≥96.0% (by total nitrogen basis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine, meso-Tetra(4-pyridyl)porphine

T2222-1G T2222-5G
TCIAT2222-1GEA 143 EUR
TCIAT2222-1G TCIAT2222-5G
5,10,15,20-Tetra(4-pyridyl)porphyrin ≥96.0% (by total nitrogen basis)
5,10,15,20-Tetra(4-pyridyl)porphyrin
Formula: C₄₀H₂₆N₈
Peso molecolare: 618,69 g/mol
MDL Number: MFCD00009626
CAS Number: 16834-13-2
EINECS: 240-858-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b089be0765c357356d9ba656faf049725b2763b&vid=2b089be0765c357356d9ba656faf049725b2763b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Gray to Dark purple to Black powder to crystal
Purity (HPLC) min. 90.0 area%
Purity (with Total Nitrogen) min. 96.0 %
Lambda max. 437.0 to 440.0 nm (0.1mol/L HCl)
Molar extinction coefficient min. 150000 (3mg/L, 0.1mol/L HCl, 437.0 to 440.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR