Stampa…

1,2,3,4-Tetra-O-acetyl-ɑ-L-fucopyranose ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 1,2,3,4-Tetra-O-acetyl-α-L(−)-fucose, Tetra-O-acetyl-α-L(−)fucopyranose, ɑ-L-Fucopyranose 1,2,3,4-Tetraacetate

T2207-1G T2207-5G
TCIAT2207-1GEA 99 EUR
TCIAT2207-1G TCIAT2207-5G
1,2,3,4-Tetra-O-acetyl-ɑ-L-fucopyranose ≥98.0% (by HPLC)
1,2,3,4-Tetra-O-acetyl-alpha-L-fucopyranose
Formula: C₁₄H₂₀O₉
Peso molecolare: 332,31 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00069791
CAS Number: 64913-16-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 92.0 to 96.0 deg-C
Specific rotation [a]20/D -115.0 to -125.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR