Stampa…

Triphenylphosphine-3,3',3''-trisulphonic acid trisodium salt ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Sodium 3,3',3''-phosphinetriyltribenzenesulphonate, Trisodium 3,3',3''-Phosphinetriyltribenzenesulphonate, Tris(3-sulphophenyl)phosphine trisodium salt, Tris(3-sulphonatophenyl)phosphine, sodium salt

T2187-1G T2187-5G
TCIAT2187-1GEA 49 EUR
TCIAT2187-1G TCIAT2187-5G
Triphenylphosphine-3,3',3''-trisulphonic acid trisodium salt ≥97.0% (by HPLC, titration analysis)
Triphenylphosphine-3,3',3''-trisulphonic acid trisodium salt
Formula: C₁₈H₁₂Na₃O₉PS₃
Peso molecolare: 568,408 g/mol
MDL Number: MFCD00145472
CAS Number: 63995-70-0
EINECS: 264-596-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Ion exchange titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR