Stampa…

2,3,4,6-Tetra-O-benzoyl-D-mannopyranose ≥93.0% (by HPLC)

Fornitore: TCI EUROPE

T2056-1G T2056-5G
TCIAT2056-1GEA 88 EUR
TCIAT2056-1G TCIAT2056-5G
2,3,4,6-Tetra-O-benzoyl-D-mannopyranose ≥93.0% (by HPLC)
2,3,4,6-Tetra-O-benzoyl-D-mannopyranose
Formula: C₃₄H₂₈O₁₀
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD02683399
CAS Number: 113544-59-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 93.0 area%
Melting point 179.0 to 185.0 deg-C
Specific rotation [a]20/D -75.0 to -80.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR