Stampa…

exo-Tetrahydrodicyclopentadiene ≥94.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: exo-Tricyclo[5.2.1.0(2,6)]decane, (3aR,4S,7R,7aS)-rel-Octahydro-1H-4,7-methanoindene

T1994-25G T1994-5G
TCIAT1994-25GEA 451 EUR
TCIAT1994-25G TCIAT1994-5G
exo-Tetrahydrodicyclopentadiene ≥94.0% (by GC)
exo-Tetrahydrodicyclopentadiene
Formula: C₁₀H₁₆
Peso molecolare: 136,24 g/mol
Flash Pt: 55°C
MDL Number: MFCD01714931
CAS Number: 2825-82-3
EINECS: 220-585-8
UN: 1993
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR