2,3,5,6-Tetrafluoro-p-xylene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,3,5,6-Tetrafluoro-1,4-dimethylbenzene
T1947-1G
T1947-5G
TCIAT1947-1GEA
60
EUR
TCIAT1947-1G
TCIAT1947-5G
2,3,5,6-Tetrafluoro-p-xylene ≥98.0% (by GC)
2,3,5,6-Tetrafluoro-p-xylene
Formula:
C₈H₆F₄ Peso molecolare: 178,13 g/mol |
MDL Number:
MFCD00012231 CAS Number: 703-87-7 UN: 1325 ADR: 4.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 32.0 to 36.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...