Stampa…

Ethyl-2-(triphenylphosphoranylidene)acetate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Ethyl triphenylphosphoranylidene-acetate, (Triphenylphosphoranylidene)acetic acid ethyl ester, (Carbethoxymethylene)triphenylphosphorane, Ethoxycarbonylmethylene-triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)acetate

T1944-25G T1944-250G
TCIAT1944-25GEA 79 EUR
TCIAT1944-25G TCIAT1944-250G
Ethyl-2-(triphenylphosphoranylidene)acetate ≥98.0% (by HPLC)
Ethyl-2-(triphenylphosphoranylidene)acetate
Formula: C₂₂H₂₁O₂P
Peso molecolare: 348,38 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00009183
CAS Number: 1099-45-2
EINECS: 214-151-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 126.0 to 130.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR