Stampa…

1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,4,8,11-Tetramethylcyclam

T1877-200MG T1877-1G
TCIAT1877-200MGEA 42 EUR
TCIAT1877-200MG TCIAT1877-1G
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane ≥98.0% (by GC, titration analysis)
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane
Formula: C₁₄H₃₂N₄
Peso molecolare: 256,44 g/mol
MDL Number: MFCD00005106
CAS Number: 41203-22-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 38.0 to 42.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR