Stampa…

1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: DOTA, 1,4,7,10-Tetraazacyclododecane-N,N',N'',N'''-tetraacetic acid, 2,2',2'',2'''-(1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetrayl)tetraacetic acid

T1875-1G T1875-200MG
TCIAT1875-1GEA 363 EUR
TCIAT1875-1G TCIAT1875-200MG
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid ≥97.0% (by titrimetric analysis)
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid
Formula: C₁₆H₂₈N₄O₈
Peso molecolare: 404,42 g/mol
Punto di fusione: 267°C
MDL Number: MFCD00068657
CAS Number: 60239-18-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (Chelometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR