3,4,5-Tris(benzyloxy)benzyl alcohol ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
3,4,5-Tribenzyloxybenzyl Alcohol, (3,4,5-Tris(benzyloxy)phenyl)methanol
T1840-5G
T1840-1G
TCIAT1840-5GEA
162
EUR
TCIAT1840-5G
TCIAT1840-1G
3,4,5-Tris(benzyloxy)benzyl alcohol ≥98.0% (by HPLC)
3,4,5-Tris(benzyloxy)benzyl alcohol
Formula:
C₂₈H₂₆O₄ Peso molecolare: 426,51 g/mol |
MDL Number:
MFCD02093503 CAS Number: 79831-88-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 105.0 to 108.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...