Stampa…

5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin ≥90.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)-21H,23H-porphine

T1815-1G T1815-100MG
TCIAT1815-1GEA 676 EUR
TCIAT1815-1G TCIAT1815-100MG
5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin ≥90.0% (by HPLC)
5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin
Formula: C₄₄H₃₀N₄O₈
Peso molecolare: 742,74 g/mol
MDL Number: MFCD02093497
CAS Number: 145764-54-1

Visualizza i codici

Risultati del test di specificazione

Appearance Dark red to Dark purple to Dark blue powder to crystal
Purity (HPLC) min. 90.0 area%
Lambda max. 415.0 to 419.0 nm (MeOH)
Absorbance (E1%1cm) min. 5000 (MeOH, 415.0 to 419.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR