Stampa…

2,2',6,6'-Tetra-tert-butyl-4,4'-dihydroxybiphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2,2',6,6'-Tetra-tert-butyl-[1,1'-biphenyl]-4,4'-diol

T1807-200MG T1807-1G
TCIAT1807-200MGEA 56 EUR
TCIAT1807-200MG TCIAT1807-1G
2,2',6,6'-Tetra-tert-butyl-4,4'-dihydroxybiphenyl ≥98.0% (by GC)
2,2',6,6'-Tetra-tert-butyl-4,4'-dihydroxybiphenyl
Formula: C₂₈H₄₂O₂
Peso molecolare: 410,64 g/mol
MDL Number: MFCD00069443
CAS Number: 128-38-1
EINECS: 204-883-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3eee0be7c9b679332d05e72362a5f6de41b4801&vid=b3eee0be7c9b679332d05e72362a5f6de41b4801&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 184.0 to 187.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR