Stampa…

Tri-O-acetyl-D-galactal ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (2R,3R,4R)-2-(Acetoxymethyl)-3,4-dihydro-2H-pyran-3,4-diyl diacetate, 1,5-Anhydro-2-deoxy-D-lyxo-hex-1-enitol 3,4,6-Tri-O-acetyl Ether, 3,4,6-Tri-O-acetyl-D-galactal

T1734-1G T1734-5G
TCIAT1734-1GEA 47 EUR
TCIAT1734-1G TCIAT1734-5G
Tri-O-acetyl-D-galactal ≥95.0% (by GC)
Tri-O-acetyl-D-galactal
Formula: C₁₂H₁₆O₇
Peso molecolare: 272,25 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00064092
CAS Number: 4098-06-0
EINECS: 223-859-5

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %
Specific rotation [a]20/D -16.0 to -21.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR