5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)porphyrin ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)-21H,23H-porphine
T1729-1G
T1729-100MG
TCIAT1729-1GEA
730
EUR
TCIAT1729-1G
TCIAT1729-100MG
5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)porphyrin ≥98.0% (by HPLC)
5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)porphyrin
Formula:
C₅₆H₅₄N₄ Peso molecolare: 783,08 g/mol |
MDL Number:
MFCD01321200 CAS Number: 56396-12-4 |
Risultati del test di specificazione
Appearance | Dark purple powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...