1-(Triisopropylsilyl)pyrrole ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-(Triisopropylsilyl)-1H-pyrrole
T1677-25ML
T1677-5ML
TCIAT1677-25MLEA
267
EUR
TCIAT1677-25ML
TCIAT1677-5ML
1-(Triisopropylsilyl)pyrrole ≥96.0% (by GC)
1-(Triisopropylsilyl)pyrrole
Formula:
C₁₃H₂₅NSi Peso molecolare: 223,44 g/mol Flash Pt: 107°C |
MDL Number:
MFCD00054932 CAS Number: 87630-35-1 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...