Stampa…

2,6,10-Trimethyl-2,6,10-triazaundecane ≥96.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine

T1618-25ML T1618-500ML
TCIAT1618-25MLEA 19 EUR
TCIAT1618-25ML TCIAT1618-500ML
2,6,10-Trimethyl-2,6,10-triazaundecane ≥96.0% (by GC, titration analysis)
2,6,10-Trimethyl-2,6,10-triazaundecane
Formula: C₁₁H₂₇N₃
Peso molecolare: 201,36 g/mol
Punto di ebollizione: 102°C
Flash Pt: 92°C
MDL Number: MFCD00126936
CAS Number: 3855-32-1
EINECS: 223-362-3
UN: 2735
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR