Stampa…

4-Hydroxy-6-(trifluoromethyl)pyrimidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

T1601-25G T1601-5G
TCIAT1601-25GEA 145 EUR
TCIAT1601-25G TCIAT1601-5G
4-Hydroxy-6-(trifluoromethyl)pyrimidine ≥98.0% (by GC, titration analysis)
4-Hydroxy-6-(trifluoromethyl)pyrimidine
Formula: C₅H₃F₃N₂O
Peso molecolare: 164,09 g/mol
MDL Number: MFCD00192530
CAS Number: 1546-78-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 169.0 to 171.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR