Stampa…

(S)-2-Methylbutyl-p-toluenesulphonate ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: p-Toluenesulphonic Acid (S)-2-Methylbutyl Ester

T1595-25G T1595-5G
TCIAT1595-25GEA 182 EUR
TCIAT1595-25G TCIAT1595-5G
(S)-2-Methylbutyl-p-toluenesulphonate ≥95.0% (by GC)
(S)-2-Methylbutyl-p-toluenesulphonate
Formula: C₁₂H₁₈O₃S
Peso molecolare: 242,33 g/mol
MDL Number: MFCD00013399
CAS Number: 38261-81-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR