Stampa…

Tris(2-cyanoethyl)amine ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3,3',3''-Nitrilotripropionitrile, 3,3',3''-Nitrilotripropanenitrile

T1577-25G T1577-5G
TCIAT1577-25GEA 305 EUR
TCIAT1577-25G TCIAT1577-5G
Tris(2-cyanoethyl)amine ≥99.0% (by titrimetric analysis)
Tris(2-cyanoethyl)amine
Formula: C₉H₁₂N₄
Peso molecolare: 176,22 g/mol
Punto di ebollizione: 140°C
MDL Number: MFCD00671575
CAS Number: 7528-78-1
UN: 2923
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 59.0 to 62.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR