Stampa…

Tetramethylammonium triacetoxyborohydride ≥85.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

T1553-25G T1553-5G
TCIAT1553-25GEA 286 EUR
TCIAT1553-25G TCIAT1553-5G
Tetramethylammonium triacetoxyborohydride ≥85.0% (by titrimetric analysis)
Tetramethylammonium triacetoxyborohydride
Formula: C₁₀H₂₂BNO₆
Peso molecolare: 263,10 g/mol
MDL Number: MFCD00012196
CAS Number: 109704-53-2
UN: 2813
ADR: 4.3,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (NBS Method) min. 85.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR