Stampa…

Tris(8-quinolinolato)aluminum ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 8-Quinolinol Aluminum Complex, Alq3, Aluminum Tris(8-hydroxyquinolinate), Aluminum triquinolin-8-olate

T1527-25G T1527-250G T1527-5G
TCIAT1527-25GEA 90 EUR
TCIAT1527-25G TCIAT1527-250G TCIAT1527-5G
Tris(8-quinolinolato)aluminum ≥98.0% (by titrimetric analysis)
Tris(8-quinolinolato)aluminum
Formula: C₂₇H₁₈AlN₃O₃
Peso molecolare: 459,44 g/mol
MDL Number: MFCD00191693
CAS Number: 2085-33-8
EINECS: 218-227-0
Merck Index: 14,00370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a56001c55a95e8c3eca44d2801c7a47cd1191850&vid=a56001c55a95e8c3eca44d2801c7a47cd1191850&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Drying loss max. 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR