Stampa…

D-Tagatose ≥98.0% (by HPLC)

Fornitore: TCI

Synonyms: (3S,4S,5R)-1,3,4,5,6-Pentahydroxyhexan-2-one

T1501-1G T1501-5G
TCIAT1501-1GEA 41 EUR
TCIAT1501-1G TCIAT1501-5G
D-Tagatose ≥98.0% (by HPLC)
D-Tagatose
Formula: C₆H₁₂O₆
Peso molecolare: 180.16 g/mol
MDL Number: MFCD00134449
CAS Number: 87-81-0
EINECS: 201-772-3
Merck Index: 14,09030
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ccc4787f6b53a852939c4fdf2587b5cc2f9b8a7&vid=6ccc4787f6b53a852939c4fdf2587b5cc2f9b8a7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Melting point 130.0 to 135.0 deg-C
Specific rotation [a]20/D -5.0 to -6.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR