Stampa…

(R)-(+)-1,1,2-Triphenyl-1,2-ethanediol ≥98.0% (by GC)

Fornitore: TCI

T1499-1G T1499-5G
TCIAT1499-1GEA 59 EUR
TCIAT1499-1G TCIAT1499-5G
(R)-(+)-1,1,2-Triphenyl-1,2-ethanediol ≥98.0% (by GC)
(R)-(+)-1,1,2-Triphenyl-1,2-ethanediol
Formula: C₂₀H₁₈O₂
MDL Number: MFCD00134424
CAS Number: 95061-46-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 130.0 deg-C
Specific rotation [a]20/D +215.0 to +225.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR