Stampa…

(1R,2S)-(-)-trans-2-Phenyl-1-cyclohexanol ≥98.0% (by GC)

Fornitore: TCI EUROPE

T1490-1G T1490-100MG
TCIAT1490-1GEA 303 EUR
TCIAT1490-1G TCIAT1490-100MG
(1R,2S)-(-)-trans-2-Phenyl-1-cyclohexanol ≥98.0% (by GC)
(1R,2S)-(-)-trans-2-Phenyl-1-cyclohexanol
Formula: C₁₂H₁₆O
Peso molecolare: 176,26 g/mol
Punto di ebollizione: 105°C
MDL Number: MFCD00075488
CAS Number: 98919-68-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C
Specific rotation [a]20/D -55.0 to -61.0 deg (C=1, MeOH)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR