Stampa…

p-Tolualdehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4'-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone

T1449-1G T1449-10G
TCIAT1449-1GEA 20 EUR
TCIAT1449-1G TCIAT1449-10G
p-Tolualdehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by titrimetric analysis)
p-Tolualdehyde 2,4-Dinitrophenylhydrazone
Formula: C₁₄H₁₂N₄O₄
Peso molecolare: 300,27 g/mol
MDL Number: MFCD00024382
CAS Number: 2571-00-8
UN: 1325
ADR: 4.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point (Decomposition) 240.0 to 245.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR