Stampa…

Copper(I) trifluoromethanesulphonate benzene complex ≥90.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Trifluoromethanesulphonic acid Copper(I) salt Benzene Complex, Copper(I) Triflate Benzene Complex

T1442-1G T1442-5G
TCIAT1442-1GEA 36 EUR
TCIAT1442-1G TCIAT1442-5G
Copper(I) trifluoromethanesulphonate benzene complex ≥90.0% (by titrimetric analysis)
Copper(I) trifluoromethanesulphonate benzene complex
Formula: [CuOSO₂CF₃]₂.C₆H₆
Peso molecolare: 503,33 g/mol
Punto di fusione: 160 °C (dec.)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00043300
CAS Number: 42152-46-5
EINECS: 255-686-6
UN: 3175
ADR: 4.1,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Potassium permanganate method) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR