Tricyclo[6.2.1.0(2,7)]undeca-4-ene ≥96.0% (by GC)
Fornitore: TCI EUROPE
T1384-25ML
T1384-500ML
TCIAT1384-25MLEA
30
EUR
TCIAT1384-25ML
TCIAT1384-500ML
Tricyclo[6.2.1.0(2,7)]undeca-4-ene ≥96.0% (by GC)
Tricyclo[6.2.1.0(2,7)]undeca-4-ene
Formula:
C₁₁H₁₆ Punto di ebollizione: 209°C |
MDL Number:
MFCD00153968 CAS Number: 91465-71-3 |
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |