(3S)-(-)-3-(Trifluoroacetamido)pyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
T1366-1G
T1366-25G
TCIAT1366-1GEA
51
EUR
TCIAT1366-1G
TCIAT1366-25G
(3S)-(-)-3-(Trifluoroacetamido)pyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)
(3S)-(-)-3-(Trifluoroacetamido)pyrrolidine hydrochloride
Formula:
C₆H₉F₃N₂O Peso molecolare: 218,60 g/mol |
MDL Number:
MFCD00143195 CAS Number: 132883-43-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 232.0 to 236.0 deg-C |
Specific rotation [a]20/D | -25.0 to -29.0 deg (C=1, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...