Polyethylene glycol dimethacrylate n=∼4 stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
PEGDMA, Polyethylene Glycol Dimethacrylate average Mn 550, PEG dimethacrylate, Polyethylene Glycol Dimethacrylate
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
T1352-25G
T1352-500G
TCIAT1352-25GEA
0
EUR
TCIAT1352-25G
TCIAT1352-500G
Polyethylene glycol dimethacrylate n=∼4 stabilizzato
Polyethylene glycol dimethacrylate
Formula:
(C₂H₄O)nC₈H₁₀O₃ Flash Pt: 192°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00014932 CAS Number: 25852-47-5 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
The average value of n | 3.0 to 5.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...